VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
Certificates
About this item
Square PETG bottles are ideal for storage of media, laboratory reagents and biological buffers up to −70 °C. Eliminate washing, depyrogenation and autoclaving steps with sterile and non-pyrogenic.
- Made from 100% virgin PETG material, durable and break resistant
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Leakproof sealing with integrated lip seal feature
- Sterile and non-pyrogenic, SAL 10⁻⁶
- Low extractables
- Transparency and reduced permeability
- Arranged in shrink-wrapped trays and polybag for prevent contamination
- 2 L product come with in mould hand grip for better ergonomics
- Products 30 ml and above has volume graduation
Certifications: US FDA 21CRF, USP CLASS VI compliant Resin.
Accessories information: Spare closures are available as per specific application requirements.
Packaging: Bottles are packed with prefer band, tray arranged and poly bagged. Spare Closures are bulk packed.
Related Information
Certificates
Search by Lot NumberCoC VWR® Media Bottles, Square, PETG COC_VWRI210-000057_296V-52-16J-110823 COC_VWRI210-000058_325V-52-16J-230823 COC_VWRI210-000060_195V-52-16J-100723 COC_VWRI210-000062_V-52-16J-140723
More
Frozen Serum Cell Growth Test Hydrogen Peroxide Permeability Test PETG Bottles Extractables and Leachable
Application Notes
VWR PETG Square Media Bottles VWR® PETG Square Media Bottles
Product Literature
Spare closures
Product Information
Frequently Bought Together