ContactUsLinkComponent
undefined | Cryogenic Box Racks
Catalog # HERA1950842
Supplier:  Thermo Scientific
undefined
Catalog # HERA1950842
Supplier:  Thermo Scientific
Specifications
  • Colour code:
    Silver
  • For:
    Thermo Scientific Matrix 200 µl Screwtop universal tubes, 500 µl storage tubes, 500 µl storage tubes with DuraSeal, 0,5 ml Nunc cryobank tubes, Thermo Scientific CryoExtra 94 (holds 92 racks)
  • W×D×H ext.:
    135×930×838 mm
  • Description:
    Rectanglular racks for tubes (cryogenic and ultra-low chest freezers), holds plates (22/rack)
  • Pk:
    1
Specifications
About this item
A convenient, streamlined process to maximize sample storage in your laboratory.
Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage are designed to hold tubes ranging from 200 up to 5,0 μl, offering maximised sample storage capacity for ultra-low chest freezers and large-capacity cryogenic freezers.