Sterilin™ Round Base Rimless Borosilicate Glass Rimless Tubes
: STERILIN
Trust these round-base borosilicate glass tubes in high temperature, aggressive chemical applications. Thermo Scientific™ Sterilin™ Round Base Rimless Borosilicate Glass Tubes are ideal for use with samples that are sensitive to plastic tube leachables. Not available in America.
- Manufactured from Pyrex borosilicate glass
- High resistance to temperature and chemical attack
- Tube profile rimless
- Supplied without caps
- Autoclavable at 121 °C
- Available in UK only
Frequently Bought Together