ContactUsLinkComponent
Shodex HILICpak VN-50 HILIC Columns for Oligonucleotides and Oligosaccharides
Shodex HILICpak VN-50 HILIC Columns for Oligonucleotides and Oligosaccharides
Supplier:  Resonac
Certificates
Shodex HILICpak VN-50 HILIC Columns for Oligonucleotides and Oligosaccharides
Supplier:  Resonac
Specifications
  • Catalog No:
  • SHOWF6711500
  • SHOWF7630600
  • SHOWF7630500
  • SHOWF6711400
  • SHOWF6830100
  • Description:
  • HILICpak VN-50G 2A
  • HILICpak VN-50 2D
  • HILICpak VN-50 4D
  • HILICpak VN-50G 4A
  • HILICpak VN-50 10E
  • Max. pressure:
  • 10 MPa (100 bar)
  • 10 MPa (100 bar)
  • 15 MPa (150 bar)
  • pH range:
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • Temperature limit:
  • 4...60 °C
  • 4...60 °C
  • 4...60 °C
  • 4...60 °C
  • 4...60 °C