ContactUsLinkComponent
Shodex HILICpak VC-50 HILIC Columns for Cationic Analytes Including Amines
Shodex HILICpak VC-50 HILIC Columns for Cationic Analytes Including Amines
Supplier:  Resonac
Certificates
Shodex HILICpak VC-50 HILIC Columns for Cationic Analytes Including Amines
Supplier:  Resonac
Specifications
  • Catalog No:
  • SHOWF6711600
  • SHOWF7630700
  • Description:
  • HILICpak VC-50G 2A
  • HILICpak VC-50 2D
  • Max. pressure:
  • 10 MPa (100 bar)
  • pH range:
  • 2 - 12
  • 2 - 12
  • Temperature limit:
  • 4...60 °C
  • 4...60 °C