Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Mag-Bind® Universal Pathogen Core Kit

Mag-Bind® Universal Pathogen Core Kit

Supplier:   OMEGA BIO-TEK
Mag-Bind® Universal Pathogen Core Kit
Supplier:  OMEGA BIO-TEK
Certificates

2 Options Available Below

Product Details & Documents

Isolate host genomic, bacterial, fungal spore, and viral DNA and viral RNA from tissue, urine, serum, and fecal samples using magnetic beads.

  • Purified DNA is suitable for PCR, restriction digestion, and hybridisation applications

The Mag-Bind® Universal Pathogen Core Kit allows rapid and reliable isolation of high-quality host genomic DNA, gram-positive and negative bacterial DNA, fungal spore DNA, viral DNA, and viral RNA from tissue, urine, serum, and fecal samples. The system incorporates a recommended enzymatic lysis which allows for fully automated extraction post-lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen Biosprint, Tecan Fluent® Automation Workstation, Tecan Freedom EVO®, and other liquid handling instruments. There are no organic extractions, thus reducing plastic waste and hands-on time and multiple samples can be processed in parallel. The system combines the Mag-Bind® technology with RBB Buffer to eliminate PCR inhibiting compounds within the samples. Purified DNA is suitable for PCR, restriction digestion, and hybridisation applications.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • OMEGM4030-00
  • OMEGM4030-01
  • Target molecule:
  • DNA
  • DNA
  • Refine target molecule:
  • Total DNA
  • Total DNA
  • Sample type:
  • Bacteria
  • Bacteria
  • Method / Format:
  • Magnetic beads
  • Magnetic beads
  • Prep size:
  • Scalable
  • Scalable

Specifications

Product Family Options

Product Information

  • Contents
    Pk
    Availability
    Price
  • OMEGM4030-00
    Universal pathogen kit
    Universal pathogen kit
    1×96 preps
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 528.00
    SFr. 528.00
    1 * 1 KIT
       SFr. 528.00

    Specifications:
    More
    Specifications
    Cat. No.
    OMEGM4030-00
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Bacteria
    Method / Format
    Magnetic beads
    Prep size
    Scalable
  • OMEGM4030-01
    Universal pathogen kit
    Universal pathogen kit
    4×96 preps
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’840.00
    SFr. 1’840.00
    1 * 1 KIT
       SFr. 1’840.00

    Specifications:
    More
    Specifications
    Cat. No.
    OMEGM4030-01
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Bacteria
    Method / Format
    Magnetic beads
    Prep size
    Scalable