PermaSleeve® High Temp Heat-Shrink Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Supplier: Brady
Certificates
About this item
The B-345 PermaSleeve® material is suitable for high temperature and low outgassing applications, or applications that require greater resistance to harsh chemicals.
- 2:1 heat shrinkable sleeves are fade-resistant and flame-retardant, and meet the requirements of NASA vacuum outgassing specification SP-R-0022A
- sleeves can be used in applications that require greater resistance to harsh chemicals such as fuels, lubricants, and high power cleaning solvents
- operating temperature range of –55 to +225 °C (–67 to +437 °F)
- matte finish
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/Bradyprinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certifications: Meets SAE-AMS-DTL-23053/18 (Class 2), SAE-AS-81531, MIL-STD-202. Tested per ASTM E 162, ASTM E 662. RoHS compliant. UL recognised.
Frequently Bought Together