ContactUsLinkComponent
Cole-Parmer® Stuart WS-200 Cabinet-Style Water Stills
Cole-Parmer® Stuart WS-200 Cabinet-Style Water Stills
Supplier:  Cole Parmer
Certificates
undefined
Cole-Parmer® Stuart WS-200 Cabinet-Style Water Stills
Supplier:  Cole Parmer
Specifications
  • Catalog No:
  • STUA99294-05
  • STUA99294-20
  • STUA99294-25
  • STUA99294-00
  • STUA99294-15
  • STUA99294-10
  • Model:
  • 99294-05
  • 99294-20
  • 99294-25
  • 99294-00
  • 99294-15
  • 99294-10
  • Distillation:
  • Single distilled
  • Single distilled
  • Single distilled
  • Single distilled
  • Double distilled
  • Double distilled
  • Output:
  • 4 L/h
  • 8 L/h
  • 8 L/h
  • 4 L/h
  • 4 L/h
  • 4 L/h
  • Resistivity range:
  • 0,5 - 1,0 MΩ-cm (resistivity threshold)
  • 0,5 - 1,0 MΩ-cm (resistivity threshold)
  • 0,5 - 1,0 MΩ-cm (resistivity threshold)
  • 0,5 - 1,0 MΩ-cm (resistivity threshold)
  • 0,7 - 1,0 MΩ-cm (resistivity threshold)
  • 0,7 - 1,0 MΩ-cm (resistivity threshold)
  • W×D×H:
  • 550×240×410 mm
  • 550×240×410 mm
  • 550×240×410 mm
  • 550×240×410 mm
  • 550×240×410 mm
  • 550×240×410 mm