Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant EN-TEV Protease (from E. coli)
Anbieter:  Abcam
Zertifikate
undefined
Human Recombinant EN-TEV Protease (from E. coli)
Anbieter:  Abcam

Technische Daten

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Enzyme name:
    Ezcut Tev Protease
  • Enzyme synonyms:
    Nuclear inclusion protein A, NIa protein
  • Purity:
    ≥95%
  • Molecular weight:
    28,6 kDa
  • Formulation:
    1 mg/ml solution in 0,1 M Tris-HCl, 0,5 M NaCl, 20% glycerol, 5 mM DTT and 0,5 mM EDTA, pH 8,0

Technische Daten

Frequently Bought Together