Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Titration beakers

Titration beakers

Supplier:   Mettler - Toledo
Titration beakers
Supplier:  Mettler - Toledo
Certificates
Titration beakers

2 Options Available Below

Product Details & Documents

Disposable titration beakers made from Polypropylene.

  • Flexible, not breakable like traditional glass lab beakers
  • Disposable after use
  • Convenient 20 ml graduations make quick measurements easier
  • Good chemical resistance for the majority of common lab applications
Product Family Options

Product Information

  • Capacity
    Graduation Interval
    Availability
    Price
  • 661-0175
    Plastic beaker (80 ml), 120 pcs.
    Plastic beaker (80 ml), 120 pcs.
    100 ml
    20 ml
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 99.80 SFr.
    99.80 SFr.
    1 * 120 ST
       99.80 SFr.

  • 661-0120
    Titration beaker, PP
    Titration beaker, PP
    100 ml
    20 ml
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 329.00 SFr.
    329.00 SFr.
    1 * 480 ST
       329.00 SFr.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.