Doypack® Stand-up Paper/Aluminium/PE Zip Seal Bags
Supplier: Flexico
Certificates
About this item
Zip seal bags designed for food contact.
- Moisture and oxygen barrier
- Ideal for dry, humid, and aqueous products
- Suitable for food contact
- Zip seal closure with sound and tactile feel
- Resealable and reusable
- Upright position for shelf display
- Bottom gusset : 2×35 mm
The multi-layer paper/aluminium/polyethylene film provides excellent preservation for bag contents. The kraft paper gives the bag an authentic and natural look. The aluminium layer ensures preservation of product freshness and gives it the best protection against humidity, UV, and oxygen. This construction also guarantee flavour conservation.
Certifications: Suitable for food contact. Certificate available upon request.
Frequently Bought Together