Order Entry
Switzerland
ContactUsLinkComponent
 
Polysorbate 20 FCC
Catalog # SPCMP1177-500MLGL
Supplier:  Spectrum Chemical
CAS Number:  
Polysorbate 20 FCC
Catalog # SPCMP1177-500MLGL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay:
    70% Min.

Specification Test Results

About this item

 :
Density:1,095
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986