Order Entry
Switzerland
Orders LinkContactUsLinkComponent
1,3-Dihydroxyacetone dimer ≥98%

1,3-Dihydroxyacetone dimer ≥98%

Irritant
Supplier:   Spectrum Chemical
1,3-Dihydroxyacetone dimer ≥98%
Supplier:  Spectrum Chemical
Certificates

2 Options Available Below

Product Details & Documents

Dihydroxyacetone, Dimer, also known as glycerone, is used as an ingredient in sunless tanning products and can be derived from the fermentation of glycerin.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
  • CAS Number:62147-49-3
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00051019
  • Molecular Weight:180.16 g/mol
  • Storage Temperature:Refrigerator
Specifications for Product Family

Specification Test Results

  • Assay:
    98% min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass, amber
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 485.00
    SFr. 485.00
    1 * 1 kg
       SFr. 485.00

  • Fiber drum
    25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.