Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

β-Alanine

Supplier:   Spectrum Chemical
β-Alanine
Supplier:  Spectrum Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - SFr. 669.00
    SFr. 669.00
    1 * 2,5 kg
       SFr. 669.00

  • Poly pail
    12 kg
    Log in for availability
    Log in for availability
    1 * 12 kg
    1 * 12 kg - SFr. 2’370.00
    SFr. 2’370.00
    1 * 12 kg
       SFr. 2’370.00

  • Fiber drum
    50 kg
    Log in for availability
    Log in for availability
    1 * 50 kg
    1 * 50 kg

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.