Order Entry
Switzerland
ContactUsLinkComponent
D-(+)-Biotin ≥97.5%, powder FCC
D-(+)-Biotin ≥97.5%, powder FCC
Supplier:  Spectrum Chemical
Certificates
D-(+)-Biotin ≥97.5%, powder FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (C₁₀H₁₆N₂O₃S):
    97.5 - 100.5%
  • Melting Range:
    229 - 232°C
  • Optical Rotation:
    +89 to +93
  • Lead (Pb):
    max. 2 mg/kg
  • Identification:
    To Pass Test

Specification Test Results

Frequently Bought Together