Sodium bisulphite, körnig FCC
:7631-90-5
:
:HNaO₃S
:MFCD00167602
:12,08731
:300 °C
:2,476 g/cm³ (20 °C)
:231-548-0
:2693
:8,III
:104.06 g/mol
:GHS05
:GHS07
:Raumtemperatur
- Assay:58.5% Min.
Frequently Bought Together
Natriumcarbonat, wasserfrei 99,5-100,5% NF, Ph. Eur., BP
: VWR Chemicals
Ammoniummetavanadat ≥99,0% analytisches Reagens
: VWR Chemicals
Natriumdodecylsulfat (SDS) ≥98%, GPR RECTAPUR®
: VWR Chemicals
Kaliumiodid ≥99,5%, AnalaR NORMAPUR® analytisches Reagens
: VWR Chemicals
Ammoniumchlorid ≥99,5% ACS
: VWR Chemicals
Calciumoxid ≥92%, Pulver, TECHNICAL
: VWR Chemicals