Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydroxide ≥95.0%, pellets FCC
Corrosive
Irritant
Sodium hydroxide ≥95.0%, pellets FCC
Catalog # SPCMS1303-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Corrosive
Irritant
Sodium hydroxide ≥95.0%, pellets FCC
Catalog # SPCMS1303-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (Total Alkali As NaOH):
    95.0 - 100.5%
  • Arsenic (As):
    max. 3 mg/kg
  • Carbonate (As Na₂CO₃):
    max. 3.0 %
  • Insoluble Substances & Organic Matter:
    To Pass Test
  • Lead (Pb):
    max. 2 mg/kg

Specification Test Results

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
UN:1823
ADR:8,II
 :GHS05
 :GHS07
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548