p-Anisaldehyde ≥98%, Reagent Grade
Catalog # SPCMAN122-10LTBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
Specifications
Specification Test Results
- Assay:98% Min.
Specification Test Results
About this item
:
Merck Index:13,00666
Boiling Pt:140 °C (1013 hPa)
Melting Pt:0…2 °C
Density:1,119
Flash Pt:110 °C
:136.15 g/mol
Storage Temperature:Ambient
EINECS:204-602-6
CAS Number:123-11-5
Formula:C₈H₈O₂
MDL Number:MFCD00003385