Spécifications
- Antibody type:Primary
- Antigen name:GPR84
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:1-100/396
- Form:Liquid
- Gene ID:53831
- Antigen synonyms:Inflammation-related G-protein coupled receptor EX33|GPCR 4|G protein coupled receptor EX 33|Inflammation related G protein coupled receptor EX33|EX 33|GPR 84|GPR84|G protein coupled receptor EX33|GPCR GPR84|G protein coupled receptor 84|GPCR EX33|GPR84_HUMAN|GPCR4|EX33|Inflammation related G protein coupled receptor EX 33|G-protein coupled receptor 84
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:44 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPR84
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Spécifications
À propos de cet article
G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates, and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. G protein receptor 84 (GPR84), a member of the GCPR 1 family, is an orphan GCPR expressed in bone marrow, brain, heart, muscle, colon, thymus, spleen, kidney, liver, placenta, intestine, lung and peripheral blood leukocytes. In activated T cells, GPR84 regulates early interleukin-4 (IL-4) gene expression.
Type: Primary
Antigen: GPR84
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/396
Host: Rabbit
Isotype: IgG
Reactivity: Mouse