Auftragserfassung
Switzerland
ContactUsLinkComponent
 
Anti-RNF15 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9160R-A680
 :  Bioss
Anti-RNF15 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9160R-A680
 :  Bioss

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    RNF15
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 680
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitop:
    201-300/465
  • Form:
    Liquid
  • Gen-ID:
    10475
  • Antigen-Synonyme:
    MGC8946|tripartite mot containing 38|RING finger protein 15|RNF15|tripartite mot-containing 38|Zinc finger protein RoRet|Gm23|RORET|RP23 480B19.4
  • Modification:
    Unmodified
  • Lagerungspuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Lagertemperatur:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Konzentration:
    1 µg/µl
  • Transporttemperatur:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human RNF15
  • Reinigung:
    Purified by Protein A
  • Packung:
    Vial
  • VE:
    100 µl

 

 

RORET, also known as RING finger protein 15 (RNF15) or zinc finger protein RoRet, is a 465 amino acid member of the TRIM family, also known as the RING-B-box coiled-coil (RBCC) family. Members of the RBCC family have an N-terminal RING finger, followed by one or two zinc-binding domains (B-box domains), a leucine coiled-coil region and a variable C-terminal domain. Found in all eukaryotes, members of the RBCC family typically function within a larger protein complex and possess ubiquitin-protein isopeptide ligase activity.The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified.

Type: Primary
Antigen: RNF15
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/465
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse