Anti-UBE2D1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-8356R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:UBE2D1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:81-147/147
- Form:Liquid
- Gene ID:7321
- Antigen synonyms:Ubiquitin conjugating enzyme E2 D1|Ubiquitin conjugating enzyme E2D 1 UBC4/5 homolog yeast|E217KB1|UBCH 5|Ubiquitin-protein ligase D1|UBCH5A|Ubiquitin-conjugating enzyme E217KB 1|UB2D1_HUMAN|UBC4/5 homolog|Ubiquitin carrier protein|Ubiquitin carrier protein D1|Ubiquitin conjugating enzyme E2 17 kDa 1|Ube2d1|UBCH5|UBC 4/5|UBC4/5|SFT|Ubiquitin-conjugating enzyme E2 D1|Stimulator of Fe transport|Ubiquitin protein ligase D1|Ubiquitin conjugating enzyme E2D 1|Ubiquitin-conjugating enzyme E2-17 kDa 1|UBCH 5A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:17 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human UBE2D1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. in vitro catalyzes 'Lys-48'-linked polyubiquitination. Mediates the selective degradation of short-lived and abnormal proteins. Functions in the E6/E6-AP-induced ubiquitination of p53/TP53. Mediates ubiquitination of PEX5 and auto-ubiquitination of CHIP, TRAF6 and TRIM63/MURF1. Ubiquitinates CHIP-associated HSP90AB1 in vitro. Lacks inherent specificity for any particular lysine residue of ubiquitin. Essential for viral activation of IRF3. Mediates polyubiquitination of CYP3A4.
Type: Primary
Antigen: UBE2D1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 81-147/147
Host: Rabbit
Isotype: IgG
Reactivity: