Anti-CDK2AP2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-7995R-A680
: Bioss
- Antibody type:Primary
- Antigen name:CDK2AP2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:75-126/126
- Form:Liquid
- Gene ID:10263
- Antigen synonyms:CDK2-associated protein 2|CDK2 associated protein 2|DOC-1R|FLJ10636|CDK2AP2|Cyclin dependent kinase 2 associated protein 2|DOC-1-related protein|CDKA2_HUMAN|p14|DOC 1 related protein|Cyclin-dependent kinase 2-associated protein 2|DOC1R|tumor suppressor deleted in oral cancer related 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:13 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CDK2AP2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This gene encodes a protein that interacts with cyclin-dependent kinase 2 associated protein 1. Pseudogenes associated with this gene are located on chromosomes 7 and 9. Alternatively spliced transcript variants have been observed for this gene.
Type: Primary
Antigen: CDK2AP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 75-126/126
Host: Rabbit
Isotype: IgG
Reactivity: