- Antibody type:Primary
- Antigen name:MAGEB2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:101-200/319
- Form:Liquid
- Gene ID:4113
- Antigen synonyms:Melanoma associated antigen B2|Smage2|Mage-b2|Mage-rs2|MAGE B2 antigen|DSS AHC critical interval MAGE superfamily 6|CT3.2|MAGE XP 2|melanoma antigen family B, 2|Smage-2 protein|Cancer/testis antigen 3.2|DAM6|Melanoma antigen, family B, 1|DSS/AHC critical interval gene, from MAGE superfamily, 6
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:35 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MAGEB2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localised in the DSS (dosage-sensitive sex reversal) critical region. It is expressed in testis and placenta, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21.
Type: Primary
Antigen: MAGEB2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/319
Host: Rabbit
Isotype: IgG
Reactivity: