Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD39L4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-CD39L4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6526R-A680
Supplier:  Bioss
Anti-CD39L4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6526R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD39L4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    331-380/428
  • Form:
    Liquid
  • Gene ID:
    957
  • Antigen synonyms:
    CD39 antigen-like 4|GDPase ENTPD5|PCPH|NTPDase 5|Nucleoside diphosphatase|CD39L4|UDPase ENTPD5|ER-UDPase|Uridine-diphosphatase ENTPD5|NTPDase-5|Guanosine-diphosphatase ENTPD5|Ectonucleoside triphosphate diphosphohydrolase 5|ENTPD5
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    47 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human ENTPD5/CD39L4
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Uridine diphosphatase (UDPase) that promotes protein N-glycosylation and ATP level regulation. UDP hydrolysis promotes protein N-glycosylation and folding in the endoplasmic reticulum, as well as elevated ATP consumption in the cytosol via an ATP hydrolysis cycle. Together with CMPK1 and AK1, constitutes an ATP hydrolysis cycle that converts ATP to AMP and results in a compensatory increase in aerobic glycolysis. The nucleotide hydrolyzing preference is GDP > IDP > UDP, but not any other nucleoside di-, mono- or triphosphates, nor thiamine pyrophosphate. Plays a key role in the AKT1-PTEN signaling pathway by promoting glycolysis in proliferating cells in response to phosphoinositide 3-kinase (PI3K) signaling.

Type: Primary
Antigen: CD39L4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 331-380/428
Host: Rabbit
Isotype: IgG
Reactivity: