Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-APOC4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-6338R-A750
 :  Bioss
Anti-APOC4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-6338R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    APOC4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    31-127/127
  • Form:
    Liquid
  • Gene ID:
    346
  • Antigen synonyms:
    APOC4_HUMAN|Apolipoprotein C IV|APOC 4|ApoC IV|APO C4|APOC4|Apo CIV
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    12 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human APOC4/Apolipoprotein CIV
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

This gene encodes a lipid-binding protein belonging to the apolipoprotein gene family. The protein is thought to play a role in lipid metabolism. Polymorphisms in this gene may influence circulating lipid levels and may be associated with coronary artery disease risk. This gene is present in a cluster with other related apolipoprotein genes on chromosome 19. Naturally occurring read-through transcription exists between this gene and the neighboring downstream apolipoprotein C-II (APOC2) gene.

Type: Primary
Antigen: APOC4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-127/127
Host: Rabbit
Isotype: IgG
Reactivity: