Anti-IMPAD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-6248R-A680
: Bioss
- Antibody type:Primary
- Antigen name:IMPAD1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:81-180/359
- Form:Liquid
- Gene ID:54928
- Antigen synonyms:IMPA3|Inositol monophosphatase domain containing protein 1|Myo inositol monophosphatase A3|IMPase 3|IMP 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IMPAD1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This gene encodes a member of the inositol monophosphatase family. The encoded protein is localised to the Golgi apparatus and catalyzes the hydrolysis of phosphoadenosine phosphate (PAP) to adenosine monophosphate (AMP). Mutations in this gene are a cause of GRAPP type chondrodysplasia with joint dislocations, and a pseudogene of this gene is located on the long arm of chromosome 1.
Type: Primary
Antigen: IMPAD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 81-180/359
Host: Rabbit
Isotype: IgG
Reactivity: