Order Entry
Switzerland
ContactUsLinkComponent
Estrogen (Mouse) ELISA Kit
Estrogen (Mouse) ELISA Kit
Catalog # ABCAAB285291-96
Supplier:  Abcam
CAS Number:  
undefined
Estrogen (Mouse) ELISA Kit
Catalog # ABCAAB285291-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Primary antibody reactivity:
    Mouse
  • Description:
    Estrogen (Mouse) ELISA kit

Specifications

About this item

An competitive ELISA kit to quantify Estrogen in mouse serum, plasma and other biological fluids.

  • Easy, convenient and time-saving method to measure the level of Estrogen in mouse serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 15,6 to 1000 pg/ml
  • Sensitivity: <9,375 pg/ml

Estrogen is the primary female sex hormone. It is responsible for the development and regulation of the female reproductive system and secondary sex characteristics. Estrogen may also refer to any substance, natural or synthetic, that mimics the effects of the natural hormone. The steroid 17β-estradiol is the most potent and prevalent endogenous estrogen, although several metabolites of estradiol also have estrogenic hormonal activity. Synthetic estrogens are used as part of some oral contraceptives, in estrogen replacement therapy for postmenopausal women, and in hormone replacement therapy for trans women. BioVision’s Estrogen ELISA kit is a Competitive ELISA assay for the quantitative measurement of mouse Estrogen in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of human Estrogen captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.