Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant Superoxide Dismutase 1 (from E. coli)
Catalog # NOVUNBC1-18544
Anbieter:  Novus Biologicals
CAS-Nummer:  
undefined
Human Recombinant Superoxide Dismutase 1 (from E. coli)
Catalog # NOVUNBC1-18544
Anbieter:  Novus Biologicals
CAS-Nummer:  

Technische Daten

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
  • Endotoxin content:
    <1,0 EU per 1 mg of protein (determined by LAL method)
  • Enzyme name:
    Superoxide Dismutase 1
  • Enzyme synonyms:
    ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110 superoxide dismutase, cystolic
  • Purity:
    >95% pure by SDS-PAGE
  • Concentration:
    1 mg/ml
  • Tested applications:
    ELISA

Technische Daten

Über diesen Artikel

The Recombinant Human SOD1/Cu-Zn SOD Protein is derived from E. coli. The Recombinant Human SOD1/Cu-Zn SOD Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

  • Entrez Gene ID: 6647

Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.