- Antibody type:Primary
- Antigen name:Cyclin M3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:251-350/707
- Form:Liquid
- Gene ID:26505
- Antigen synonyms:DKFZp434I1016|Metal transporter CNNM3|Ancient conserved domain containing protein 3|FLJ20018|CNNM3_HUMAN|ACDP3|CNNM3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:76 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Cyclin M3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Cyclin M3 is a 707 amino acid multi-pass membrane protein that shares weak sequence similarity with cyclin proteins, yet displays no cyclin-like function in vivo. Though ubiquitously expressed, Cyclin M3 is found at highest levels in kidney, brain, spleen and heart. Cyclin M3 is localised to the nucleus where it is likely a metal transporter. Cyclin M3 contains two CBS domains, which appear to bind ligands with an adenosyl group such as AMP, ATP and S-AdoMet and may play a regulatory role in sensitizing proteins to adenosyl-carrying ligands. There are three isoforms of Cyclin M3 that are produced as a result of alternative splicing events.
Type: Primary
Antigen: Cyclin M3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/707
Host: Rabbit
Isotype: IgG
Reactivity: