Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-SOX8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-11600R-A750
 :  Bioss
Anti-SOX8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-11600R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    SOX8
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    101-200/446
  • Form:
    Liquid
  • Antigen synonyms:
    Transcription factor SOX8|SRY box containing gene 8|SOX8_HUMAN|Transcription factor SOX 8|SRY sex determining region Y box 8|Transcription factor SOX-8|SRY box 8|Sox8|SOX 8
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    47 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SOX8
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.

Type: Primary
Antigen: SOX8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/446
Host: Rabbit
Isotype: IgG
Reactivity: Mouse