Anti-HINT1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Katalognr. BOSSBS-10228R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HINT1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Epitope:41-126/126
- Form:Liquid
- Antigen synonyms:histidine triad nucleotide binding protein 1|PKCI1|Protein kinase C inhibitor 1|Adenosine 5'-monophosphoramidase|PKCI 1|Protein kinase C-interacting protein 1|Adenosine 5' monophosphoramidase|HINT 1|Histidine triad nucleotide-binding protein 1|Protein kinase C interacting protein 1|HINT-1|HINT1_HUMAN|PRKCNH1|PKCI-1|histidine triad nucleotide binding protein|histidine triad nucleotide-binding protein
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:14 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HINT1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene can hydrolyse substrates such as AMP-morpholidate, AMP-N-alanine methyl ester, AMP-alpha-acetyl lysine methyl ester, and AMP-NH2. The encoded protein interacts with these substrates via a histidine triad motif, which is part of the loop that binds to the substrate. This gene has been found to be a tumour suppressing gene. Several transcript variants, but only one of them protein-coding, have been found for this gene.
Type: Primary
Antigen: HINT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 41-126/126
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse