Order Entry
Switzerland
ContactUsLinkComponent
Anti-RSV Nucleoprotein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-RSV Nucleoprotein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10207R-A680
Supplier:  Bioss
Anti-RSV Nucleoprotein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10207R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RSV Nucleoprotein
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Epitope:
    101-200/391
  • Form:
    Liquid
  • Antigen synonyms:
    Nucleoprotein|NCAP_HRSVA|N|Respiratory syncytial virus nucleoprotein|Nucleocapsid protein|Protein N
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    43 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Respiratory Syncytial Virus Nucleoprotein
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Respiratory syncytial virus (RSV) is a major cause of respiratory illness in young children. RSV infection produces a variety of signs and symptoms involving different areas of the respiratory tract, from the nose to the lungs. RSV is a negative sense, enveloped RNA virus. The virion is variable in shape and size with average diameter of between 120 and 300 nm. The Nucleoprotein encapsidates the genome, protecting it from nucleases. The nucleocapsid (NC) has a helical structure. The encapsidated genomic RNA is termed the NC and serves as template for transcription and replication. During replication, encapsidation by protein N is coupled to RNA synthesis and all replicative products are resistant to nucleases.

Type: Primary
Antigen: RSV Nucleoprotein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/391
Host: Rabbit
Isotype: IgG
Reactivity: Virus