Anti-HADHSC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-10020R-A750
: Bioss
- Antibody type:Primary
- Antigen name:HADHSC
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:241-314/314
- Form:Liquid
- Gene ID:3033
- Antigen synonyms:Hydroxyacyl-coenzyme A dehydrogenase, mitochondrial|HADH1|Medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase|HAD|HADH|HADHSC|SCHAD|MSCHAD|HHF4|Short-chain 3-hydroxyacyl-CoA dehydrogenase|HCDH
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SCHAD/HADHSC
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Plays an essential role in the mitochondrial beta-oxidation of short chain fatty acids. Exerts it highest activity toward 3-hydroxybutyryl-CoA.
Type: Primary
Antigen: HADHSC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 241-314/314
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat