Order Entry
Switzerland
ContactUsLinkComponent
Anti-AMPD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-AMPD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2077R-A680
Supplier:  Bioss
Anti-AMPD1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2077R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AMPD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Epitope:
    101-200/745
  • Form:
    Liquid
  • Gene ID:
    229665
  • Antigen synonyms:
    Ampd1|Myoadenylate deaminase|Ampd-1|AMP deaminase isoform M|AI553520|AMP deaminase 1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    86 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from mouse AMP deaminase 1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

AMP deaminase plays a critical role in energy metabolism.

Type: Primary
Antigen: AMPD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/745
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat