Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-Annexin VI Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Katalognr. BOSSBS-1471R-A555
Anbieter:  Bioss
Anti-Annexin VI Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Katalognr. BOSSBS-1471R-A555
Anbieter:  Bioss

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Annexin VI
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 555
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Rat
  • Epitop:
    551-673/673
  • Form:
    Liquid
  • Antigen-Synonyme:
    Protein III|Calelectrin|CBP68|CPB II|AnnexinA6|Lipocortin VI|AnnexinVI|P70|Calphobindin II|ProteinIII.|ANX A6|Calcium binding protein p68|ANXA6|ANX6|ANX 6|Annexin VI p68|p68|LipocortinVI|67 kDa calelectrin|Annexin A6|CalphobindinII|CBP 68|ANXA 6|Chromobindin 20|Annexin VI|CPBII|Chromobindin20
  • Modification:
    Unmodified
  • Lagerungspuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    67-74 kDa
  • Lagertemperatur:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Konzentration:
    1 µg/µl
  • Transporttemperatur:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from rat Annexin VI
  • Validierte Anwendungen:
    ICC
  • Reinigung:
    Purified by Protein A
  • Packung:
    Vial
  • VE:
    100 µl

Technische Daten

Über diesen Artikel

The Annexin family of calcium binding proteins is composed of at least ten mammalian genes. It is characterised by a conserved core domain which binds to phospholipids in a Ca2+ dependent manner and a unique amino terminal region which may confer binding specificity. The Annexin family has been implicated as regulators of such diverse processes as ion flux, endocytosis and exocytosis, and cellular adhesion. When overexpressed in A431 cells, Annexin VI causes a partial reversal of the transformed phenotype.

Type: Primary
Antigen: Annexin VI
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 551-673/673
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Others