Anti-Neuroligin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4212R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Neuroligin 1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:701-800/863
- Form:Liquid
- Gene ID:22871
- Antigen synonyms:NLGN1|KIAA1070|NL1|Neuroligin-1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:87 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Neuroligin 1
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Cell surface protein involved in cell-cell-interactions via its interactions with neurexin family members. Plays a role in synapse function and synaptic signal transmission, and probably mediates its effects by recruiting and clustering other synaptic proteins. May promote the initial formation of synapses, but is not essential for this. In vitro, triggers the de novo formation of presynaptic structures. May be involved in specification of excitatory synapses (By similarity).
Type: Primary
Antigen: Neuroligin 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 701-800/863
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat