Order Entry
Switzerland
ContactUsLinkComponent
Anti-Selenium Binding Protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-Selenium Binding Protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4200R-A680
Supplier:  Bioss
Anti-Selenium Binding Protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4200R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Selenium Binding Protein 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    401-472/472
  • Form:
    Liquid
  • Gene ID:
    1954
  • Antigen synonyms:
    Selenium binding protein1|SP56|SBP|Selenbp2|SELNBP1|Selenium binding protein 2|SBP1|56 kDa selenium binding protein
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    52 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SBP1/Selenium Binding Protein 1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Selenium is an essential trace element that confers tolerance to toxicity arising through exposure to heavy metals or other reactive xenobiotics. Selenium exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. Both effects are attributed to selenium-binding proteins. Selenium binding protein 1 is down-regulated in lung adenocarcinoma, colourectal cander and ovarian cancer. It is two-fold upregulated in the brains of patients suffering from schizophrenia, and is therefore a biomarker for this disease.

Type: Primary
Antigen: Selenium Binding Protein 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-472/472
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat