Anti-ADCY7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3924R-A750
Anbieter: Bioss
Technische Daten
- Antibody type:Primary
- Antigen name:ADCY7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:901-1080/1080
- Form:Liquid
- Gene ID:113
- Antigen synonyms:Adenylyl cyclase 7|Adenylate cyclase type 7|ATP pyrophosphate-lyase 7|AC7|KIAA37|ADCY7|Adenylate cyclase type VII
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:120 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ADCY7
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Technische Daten
Über diesen Artikel
This gene encodes a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterised by the presence of twelve membrane-spanning domains in its sequences.
Type: Primary
Antigen: ADCY7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 901-1080/1080
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat