Order Entry
Switzerland
ContactUsLinkComponent
Anti-NPM Thr95 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-NPM Thr95 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-3309R-A680
Supplier:  Bioss
Anti-NPM Thr95 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-3309R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NPM Thr95
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Gene ID:
    4869
  • Antigen synonyms:
    NPM|Numatrin|Nucleophosmin/nucleoplasmin family member 1|MGC104254|NO38|Nucleophosmin phospho Thr95|OTTHUMP00000161024|Nucleolar Phosphoprotein B23|NPM_HUMAN|Nucleophosmin phospho T95|OTTHUMP00000223398|OTTHUMP00000161025|TRK fused gene|Nucleophosmin nucleolar phosphoprotein B23 numatrin|Nucleophosmin|Nucleolar protein NO38|Nucleophosmin/B23.2|OTTHUMP00000223397|B23|Nucleoplasmin Family Member 1|NMP1|p-Nucleophosmin Thr95|NPM1|NPM 1
  • Modification:
    Phosphorylation (Thr95)
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    38 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human NPM around the phosphorylation site of Thr95
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

This gene encodes a phosphoprotein which moves between the nucleus and the cytoplasm. The gene product is thought to be involved in several processes including regulation of the ARF/p53 pathway. A number of genes are fusion partners have been characterised, in particular the anaplastic lymphoma kinase gene on chromosome 2. Mutations in this gene are associated with acute myeloid leukaemia. More than a dozen pseudogenes of this gene have been identified. Alternative splicing results in multiple transcript variants.

Type: Primary
Antigen: NPM Thr95
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse