Anti-Eppin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-2365R-A750
: Bioss
- Antibody type:Primary
- Antigen name:Eppin
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:23-134/134
- Form:Liquid
- Gene ID:75526
- Antigen synonyms:SPINLW1|EPPI_MOUSE|Cancer/testis antigen 71|Variant|CT71|Serine protease inhibitor-like with Kunitz and WAP domains 1|WAP7|WFDC7|Protease inhibitor WAP7|Epididymal protease inhibitor|WAPfour-disulfide core domain protein 7
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:13 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse Eppin
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This gene encodes an epididymal protease inhibitor, which contains both kunitz-type and WAP-type four-disulfide core (WFDC) protease inhibitor consensus sequences. Most WFDC genes are localised to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene is a member of the WFDC gene family and belongs to the telomeric cluster. The protein can inhibit human sperm motility, and polymorphisms in this gene are associated with male infertility. Read-through transcription also exists between this gene and the downstream WFDC6 (WAP four-disulfide core domain 6) gene.
Type: Primary
Antigen: Eppin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 23-134/134
Host: Rabbit
Isotype: IgG
Reactivity: