Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cells
Dicer Knockout Mouse Embryonic Stem Cells Cell Lysate (Embryonic Normal)
Dicer Knockout Mouse Embryonic Stem Cells Cell Lysate (Embryonic Normal)
Dicer Knockout Mouse Embryonic Stem Cells Cell Lysate (Embryonic Normal)
Catalog # NOVUNBP1-97435
Supplier:
Novus Biologicals
CAS Number:
Expand image
Dicer Knockout Mouse Embryonic Stem Cells Cell Lysate (Embryonic Normal)
Catalog # NOVUNBP1-97435
Supplier:
Novus Biologicals
CAS Number:
Specifications
Cell Format:
Cryopreserved
Pk:
0,25 mg
Tested applications:
WB, ImmunoPrecipitation
Specifications
About this item
Product Details
Product Details