Anti-BCHE Mouse Monoclonal Antibody [clone: 1E8] (ALEXA FLUOR® 750)
Catalog # BOSSBSM-2011M-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:BCHE
- Clonality:Monoclonal
- Clone:1E8
- Conjugation:Alexa Fluor® 750
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:590
- Antigen synonyms:Acylcholine acylhydrolase|CHE1|butyrylcholinesterase,CHLE_HUMAN|Cholinesterase|Choline esterase II|Butyrylcholine esterase|Pseudocholinesterase
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:Full length native protein purified from human
- Purification:Purified by Protein G
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage.
Type: Primary
Antigen: BCHE
Clonality: Monoclonal
Clone: 1E8
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Human