Order Entry
Switzerland
ContactUsLinkComponent
Anti-BPNT1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Anti-BPNT1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-20136R-A680
Supplier:  Bioss
Anti-BPNT1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-20136R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BPNT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    2-100/308
  • Form:
    Liquid
  • Gene ID:
    10380
  • Antigen synonyms:
    3'(2'),5'-bisphosphate nucleotidase 1|PIP|BPNT1|Bisphosphate 3'-nucleotidase 1|PAP-inositol 1,4-phosphatase|HEL20
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    33 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human BPNT1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Converts adenosine 3'-phosphate 5'-phosphosulfate (PAPS) to adenosine 5'-phosphosulfate (APS) and 3'(2')-phosphoadenosine 5'- phosphate (PAP) to AMP. Has 1000-fold lower activity towards inositol 1,4-bisphosphate (Ins(1,4)P2) and inositol 1,3,4-trisphosphate (Ins(1,3,4)P3), but does not hydrolyse Ins(1)P, Ins(3,4)P2, Ins(1,3,4,5)P4 or InsP6.

Type: Primary
Antigen: BPNT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 2-100/308
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat