Specifications
- Antibody type:Primary
- Antigen name:IGLL1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:151-213/213
- Form:Liquid
- Antigen synonyms:AGM2|Immunoglobulin omega polypeptide|CD179b antigen|VPREB2|IGL1|Ig lambda-5|Immunoglobulin-related protein 14,1|IGO|Ig lambda 5|CD179 antigen-like family member B|IGVPB|Immunoglobulin lambda-like polypeptide 1|Lambda5|IGLJ14,1|Igll1|Pre B lymphocyte specic protein 2|IGLL1_HUMAN|Immunoglobulin related protein 14,1|IGLL|CD179B|Immunoglobulin lambda like polypeptide 1 precursor|Immunoglobulin omega polypeptide chain|IGL5|14,1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:19 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IGLL1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The preB cell receptor is found on the surface of proB and preB cells, where it is involved in transduction of signals for cellular proliferation, differentiation from the proB cell to the preB cell stage, allelic exclusion at the Ig heavy chain gene locus, and promotion of Ig light chain gene rearrangements. The preB cell receptor is composed of a membrane-bound Ig mu heavy chain in association with a heterodimeric surrogate light chain. This gene encodes one of the surrogate light chain subunits and is a member of the immunoglobulin gene superfamily. This gene does not undergo rearrangement. Mutations in this gene can result in B cell deficiency and agammaglobulinemia, an autosomal recessive disease in which few or no gamma globulins or are made. Two transcript variants encoding different isoforms have been found for this gene.
Type: Primary
Antigen: IGLL1
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 151-213/213
Host: Rabbit
Isotype: IgG
Reactivity: Mouse