Anti-ITM5 Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15518R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ITM5
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:1-100/132
- Form:Liquid
- Antigen synonyms:Interferon-induced transmembrane protein 5|Hrmp1|M5_HUMAN|BRIL|Bone-restricted interferon-induced transmembrane protein-like protein|Fragilis4|ITM5
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:14 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IFITM5
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
IFITM5 is a membrane protein thought to play a role in bone mineralisation. This gene is located on chromosome 11 in a cluster of related genes which are induced by interferon, however, this gene has not been shown to be interferon inducible. A similar gene, located in a gene cluster on mouse chromosome 7, is a member of the interferon-inducible fragilis gene family. The mouse gene encodes a transmembrane protein described as participating in germ cell competence. A mutation in the 5' UTR of this gene has been associated with osteogenesis imperfecta type V (PMID: 22863190, 22863195).
Type: Primary
Antigen: ITM5
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 1-100/132
Host: Rabbit
Isotype: IgG
Reactivity: