- Pk:96 Tests
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:S100B ELISA kit
A sandwich ELISA kit for the quantitative measurement of S100B in mouse serum, plasma and other biological fluids.
- Detection range: 78,13 to 5000 pg/ml
- Precision: coefficient of variation is <10%
- Quantitative measurement of S100B in mouse serum, plasma and other biological fluids
- Sensitivity: 46,88 pg/ml
- Specificity: This kit recognises mouse S100B in samples
- No significant cross-reactivity or interference between mouse S100B and analogues was observed
S100 proteins regulate a variety of cellular processes such as cell growth and motility, cell cycle progression, transcription, and differentiation. Research studies have demonstrated that significant correlation exists between aberrant S100 protein expression and cancer progression. S100 proteins primarily mediate immune responses in various tissue types but are also involved in neuronal development. S100B (mouse) ELISA kit is based on sandwich-ELISA principle. The micro ELISA plate provided in this kit has been pre-coated with an antibody specific to mouse S100B. Standards or samples are added to the micro ELISA plate wells that bind to the specific antibody. Then a biotinylated detection antibody specific for mouse S100B and Avidin-Horseradish Peroxidase (HRP) conjugate are added successively to each micro plate well and incubated. The wells are washed, a TMB substrate solution is added to the wells and blue color develops in proportion to the amount of S100B bound. The enzyme-substrate reaction is terminated by the addition of stop solution and the color turns yellow. The optical density (OD) is measured spectrophotometrically at a wavelength of 450 ±2 nm. The OD value is proportional to the concentration of mouse S100B. The concentration of mouse S100B in the samples can be calculated by comparing the OD of the samples to the standard curve.