About this item
RNA from TRIzol® in seven minutes.
- Extract RNA from any sample (cells, tissues, blood, biological liquids, etc.)
- Easy Handling: No phase separation or precipitation steps
- NGS-ready: Ultra-pure RNA without phenol carryover, no DNA contamination (DNase I included)
- Total RNA Recovery: All RNA >17 nt is recovered
The Direct-zol™ RNA Kits provide a streamlined method for the purification of up to 100 µg (per prep) of high-quality RNA directly from samples in TRI Reagent® or similar. Total RNA, including small RNAs (17 to 200 nt), is effectively isolated from a variety of sample sources (cells, tissues, serum, plasma, blood, biological liquids, etc.).
The procedure is easy. Simply apply a prepared sample in TRI Reagent® directly to the Zymo-Spin™ column and then bind, wash, and elute the RNA. No phase separation, precipitation, or post-purification steps are necessary. The eluted RNA is high quality and suitable for subsequent molecular manipulation and analysis (including RT-PCR, transcription profiling, hybridisation, sequencing and more).
Specifications
- Method / Format:Column (silica-based)
- Prep size:Micro
- Refine target molecule:Total RNA
- Sample type:BloodBiofluids (serum, plasma, saliva)Cultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Sample size:≤1×10⁶ cells
- Description:Direct-zol™ RNA Microprep kit
- Sample capacity:10 µg