RBC Lysis Buffer (10X)
Catalog # TNB-4300-L100
Supplier: Tonbo Biosciences
CAS Number:
Specifications
- VE:100 mL
- Lagertemperatur:Store Undiluted at 2...8 °C
Specifications
About this item
RBC Lysis Buffer (10X) is a concentrated ammonium chloride-based lysing reagent.
- Reactivity: Mouse, Human
The diluted 1X working solution will lyse red blood cells in single cell suspensions with minimal effects on leukocytes. RBC Lysis Buffer (10X) does not contain a fixative so the cells remain viable after red blood cell lysis.