Order Entry
Switzerland
ContactUsLinkComponent
PicoKine Mouse IDS/Iduronate 2 Sulfatase; ELISA Kit
PicoKine Mouse IDS/Iduronate 2 Sulfatase; ELISA Kit
Catalog # BSBTEK1453
Supplier:  Boster Bio
CAS Number:  
undefined
PicoKine Mouse IDS/Iduronate 2 Sulfatase; ELISA Kit
Catalog # BSBTEK1453
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse IDS/Iduronate 2 Sulfatase PicoKine; ELISA kit
  • Tests per kit:
    96 wells

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of mouse IDS.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect Mouse Iduronate 2-Sulfatase/Ids with <15 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Iduronate 2-Sulfatase/Ids tissue specificity: Found to be expressed in alpha and beta pancreatic cells. .

96wells/kit, with removable strips.