Order Entry
Switzerland
ContactUsLinkComponent
PicoKine Rat BMP-7; ELISA Kit
PicoKine Rat BMP-7; ELISA Kit
Catalog # BSBTEK1444
Supplier:  Boster Bio
CAS Number:  
undefined
PicoKine Rat BMP-7; ELISA Kit
Catalog # BSBTEK1444
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Description:
    Rat BMP-7 PicoKine; ELISA kit
  • Tests per kit:
    96 wells

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of rat BMP-7.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect Rat Bmp7 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: bone tissue, cell culture supernates, serum and plasma((heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Bmp7 tissue specificity:

96wells/kit, with removable strips.